accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOOOONOO

CAS RN 1611477-45-2

IUPAC Name: 4-{4'-[2-(2-{2-[2-(4-nitrophenoxy)ethoxy]ethoxy}ethoxy)ethoxy]-[1,1'-biphenyl]-4-yl}phenol
Synonyms: 4-{4'-[2-(2-{2-[2-(4-nitrophenoxy)ethoxy]ethoxy}ethoxy)ethoxy]-[1,1'-biphenyl]-4-yl}phenol
Molecular Weight (g/mol): 559.62
Molecular Formula: C32H33NO8
InChi Key: HCDBZEWQACQBAN-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=C(OCCOCCOCCOCCOC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Grade 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results