accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOCH3NNOH3CClH3CNO

CAS RN 1613695-14-9

IUPAC Name: 2-[2-(3-chloro-4-methoxyphenyl)ethyl]-5-(3,5-dimethyl-1,2-oxazol-4-yl)-1-[2-(morpholin-4-yl)propyl]-1H-1,3-benzodiazole
Synonyms: 2-[2-(3-chloro-4-methoxyphenyl)ethyl]-5-(3,5-dimethyl-1,2-oxazol-4-yl)-1-[2-(morpholin-4-yl)propyl]-1,3-benzodiazole
Molecular Weight (g/mol): 509.05
Molecular Formula: C28H33ClN4O3
InChi Key: GEPYBHCJBORHCE-UHFFFAOYNA-N
SMILES: COC1=C(Cl)C=C(CCC2=NC3=CC(=CC=C3N2CC(C)N2CCOCC2)C2=C(C)ON=C2C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results