accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPONNNOCH3

CAS RN 161583-75-1

IUPAC Name: diethyl [6-(pyridin-2-yl)-[2,2'-bipyridin]-4-yl]phosphonate
Synonyms: diethyl 6-(pyridin-2-yl)-[2,2'-bipyridin]-4-ylphosphonate
Molecular Weight (g/mol): 369.36
Molecular Formula: C19H20N3O3P
InChi Key: MDIMWBGOUZAXEP-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results