accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOPOHONOOHNNOFFF

CAS RN 161605-73-8

IUPAC Name: {[7-(morpholin-4-yl)-2,3-dioxo-6-(trifluoromethyl)-1,2,3,4-tetrahydroquinoxalin-1-yl]methyl}phosphonic acid
Synonyms: fanapanel
Molecular Weight (g/mol): 409.26
Molecular Formula: C14H15F3N3O6P
InChi Key: WZMQMKNCWDCCMT-UHFFFAOYSA-N
SMILES: OP(O)(=O)CN1C(=O)C(=O)NC2=CC(=C(C=C12)N1CCOCC1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)