accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHCH3

CAS RN 16165-33-6

IUPAC Name: diundecylamine
Synonyms: diundecylamine
Molecular Weight (g/mol): 325.63
Molecular Formula: C22H47N
InChi Key: NKGSHSILLGXYDW-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCNCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results