accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2HNONNClH2NNH2HCl(E/Z)

CAS RN 161804-20-2

IUPAC Name: 3,5-diamino-N-(N'-benzylcarbamimidoyl)-6-chloropyrazine-2-carboxamide hydrochloride
Synonyms: benzamil
Molecular Weight (g/mol): 356.21
Molecular Formula: C13H15Cl2N7O
InChi Key: ZNWMRWWNJBXNKJ-UHFFFAOYSA-N
SMILES: Cl.NC(NC(=O)C1=C(N)N=C(N)C(Cl)=N1)=NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)