CAS RN 1620-30-0
CAS RN 1620-30-0
IUPAC Name:
diphenyl(pyridin-4-yl)methanol
Synonyms:
diphenyl(pyridin-4-yl)methanol
Molecular Weight (g/mol):
261.32
Molecular Formula:
C18H15NO
InChi Key:
MRHLFZXYRABVOZ-UHFFFAOYSA-N
SMILES:
OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
alpha,alpha-Diphenyl-4-pyridylmethanol 98.0+%, TCI America™
CAS: 1620-30-0 Molecular Formula: C18H15NO Molecular Weight (g/mol): 261.32 MDL Number: MFCD00006434 InChI Key: MRHLFZXYRABVOZ-UHFFFAOYSA-N PubChem CID: 74177 IUPAC Name: diphenyl(pyridin-4-yl)methanol SMILES: OC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=NC=C1