accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOONNOOOCH3

CAS RN 162042-44-6

IUPAC Name: 1-(1-{4-[(1-acetylpiperidin-4-yl)oxy]-2-methoxybenzoyl}piperidin-4-yl)-2,4-dihydro-1H-3,1-benzoxazin-2-one
Synonyms: 1-(1-{4-[(1-acetylpiperidin-4-yl)oxy]-2-methoxybenzoyl}piperidin-4-yl)-4H-3,1-benzoxazin-2-one
Molecular Weight (g/mol): 507.59
Molecular Formula: C28H33N3O6
InChi Key: WDERJSQJYIJOPD-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(OC2CCN(CC2)C(C)=O)=C1)C(=O)N1CCC(CC1)N1C(=O)OCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)