accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNN(E/Z)(E/Z)

CAS RN 1622-12-4

IUPAC Name: 4-methyl-N'-(phenylamino)-N-(phenylimino)benzene-1-carboximidamide
Synonyms: 4-methyl-N'-(phenylamino)-N-(phenylimino)benzenecarboximidamide
Molecular Weight (g/mol): 314.39
Molecular Formula: C20H18N4
InChi Key: FEYZMOAOMKSFOX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=NNC1=CC=CC=C1)N=NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results