accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1622008-73-4

IUPAC Name: 4-(7-{4-[bis(4-methoxyphenyl)amino]phenyl}-2H,3H-thieno[3,4-b][1,4]dioxin-5-yl)-N,N-bis(4-methoxyphenyl)aniline
Synonyms: 4-(7-{4-[bis(4-methoxyphenyl)amino]phenyl}-2H,3H-thieno[3,4-b][1,4]dioxin-5-yl)-N,N-bis(4-methoxyphenyl)aniline
Molecular Weight (g/mol): 748.89
Molecular Formula: C46H40N2O6S
InChi Key: JOJPNOVYPGLCMM-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(C=C1)C1=C2OCCOC2=C(S1)C1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results