accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1623-05-8

IUPAC Name: 1,1,1,2,2,3,3-heptafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propane
Synonyms: 1,1,1,2,2,3,3-heptafluoro-3-[(1,2,2-trifluoroethenyl)oxy]propane
Molecular Weight (g/mol): 266.04
Molecular Formula: C5F10O
InChi Key: KHXKESCWFMPTFT-UHFFFAOYSA-N
SMILES: FC(F)=C(F)OC(F)(F)C(F)(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results