Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoropropoxyethylene 98.0+%, TCI America™
SDP

Catalog No. P122410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P122410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P122410G Supplier TCI America Supplier No. P122410G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1623-05-8
Molecular Formula C5F10O
Molecular Weight (g/mol) 266.038
MDL Number MFCD00191577
InChI Key KHXKESCWFMPTFT-UHFFFAOYSA-N
Synonym Perfluoro(propyl Vinyl Ether)
PubChem CID 15388
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-3-(1,2,2-trifluoroethenoxy)propane
SMILES C(=C(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F

Specifications

Color Yellow
Boiling Point 36°C
Quantity 10 g
Formula Weight 266.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Perfluoropropoxyethylene
EINECSNumber (2)-3068
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.