accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163080-99-7

IUPAC Name: 1-[(benzyloxy)carbonyl]-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 3-[(benzyloxy)carbonyl]-1-methylimidazol-1-ium triflate
Molecular Weight (g/mol): 366.31
Molecular Formula: C13H13F3N2O5S
InChi Key: ICFSDLPZJDDPHP-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.C[N+]1=CN(C=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results