Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Carbobenzoxy-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. C24205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24205G Supplier TCI America Supplier No. C24205G

missing translation for 'validMainframeMsg'

A laboratory reagent for research applications.

Chemical Identifiers

CAS 163080-99-7
Molecular Formula C13H13F3N2O5S
Molecular Weight (g/mol) 366.311
InChI Key ICFSDLPZJDDPHP-UHFFFAOYSA-M
Synonym 1-Carbobenzoxy-3-methylimidazolium Triflate, 1-Cbz-3-methylimidazolium Trifluoromethanesulfonate, 1-Cbz-3-methylimidazolium Triflate
PubChem CID 11858599
IUPAC Name benzyl 3-methylimidazol-3-ium-1-carboxylate;trifluoromethanesulfonate
SMILES C[N+]1=CN(C=C1)C(=O)OCC2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color White
Quantity 5 g
Formula Weight 366.31
Percent Purity ≥98.0% (HPLC,N)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 1-Carbobenzoxy-3-methylimidazolium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.