accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163105-90-6

IUPAC Name: (2-methoxypyridin-3-yl)boronic acid
Synonyms: 2-methoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 152.94
Molecular Formula: C6H8BNO3
InChi Key: NVOLYUXUHWBCRJ-UHFFFAOYSA-N
SMILES: COC1=NC=CC=C1B(O)O
Molecular Weight (g/mol) 
  • (5)
  • (1)
  • (1)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)