CAS RN 163163-24-4
IUPAC Name:
N-(6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-2H-1-benzopyran-4-yl)-N-methylethane-1-sulfonamide
Synonyms:
N-(6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl)-N-methylethanesulfonamide
Molecular Weight (g/mol):
324.4
Molecular Formula:
C15H20N2O4S
InChi Key:
HVSJHHXUORMCGK-UHFFFAOYNA-N
SMILES:
CCS(=O)(=O)N(C)C1C(O)C(C)(C)OC2=C1C=C(C=C2)C#N
1
–
1
of
1
results
1
R&D Systems™ (-)-[3R,4S]-Chromanol 293B
CAS: 163163-24-4 Molecular Formula: C15H20N2O4S