accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 163239-22-3

IUPAC Name: 6-fluoro-3-[2-(pyridin-3-yl)ethenyl]-1H-indole
Synonyms: 6-fluoro-3-[2-(pyridin-3-yl)ethenyl]-1H-indole
Molecular Weight (g/mol): 238.27
Molecular Formula: C15H11FN2
InChi Key: YBSDQTBCNYWBMX-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(C=CC3=CN=CC=C3)=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results