accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFFFFFFFFF

CAS RN 163702-07-6

IUPAC Name: 1,1,1,2,2,3,3,4,4-nonafluoro-4-methoxybutane
Synonyms: methyl perfluorobutyl ether
Molecular Weight (g/mol): 250.06
Molecular Formula: C5H3F9O
InChi Key: OKIYQFLILPKULA-UHFFFAOYSA-N
SMILES: COC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)