Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Nonafluorobutyl Ether 98.0+%, TCI America™
SDP

Catalog No. M1345500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1345500G 500 g
M134525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1345500G Supplier TCI America Supplier No. M1345500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 163702-07-6
Molecular Formula C5H3F9O
Molecular Weight (g/mol) 250.064
MDL Number MFCD00674093
InChI Key OKIYQFLILPKULA-UHFFFAOYSA-N
Synonym Methyl Perfluorobutyl Ether, Nonafluorobutyl Methyl Ether, Perfluorobutyl Methyl Ether
PubChem CID 164514
IUPAC Name 1,1,1,2,2,3,3,4,4-nonafluoro-4-methoxybutane
SMILES COC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Quantity 500 g
Formula Weight 250.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Nonafluorobutyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.