accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16382-18-6

IUPAC Name: ethyl 5,6-dimethoxy-1H-indole-2-carboxylate
Synonyms: ethyl 5,6-dimethoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 249.27
Molecular Formula: C13H15NO4
InChi Key: RBUSDPZPBBBAKR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(OC)=C(OC)C=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results