Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5,6-dimethoxyindole-2-carboxylate, 98+%, Thermo Scientific™

Catalog No. AAL0076203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0076203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0076203 Supplier Thermo Scientific Chemicals Supplier No. L0076203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16382-18-6
Molecular Formula C13H15NO4
Molecular Weight (g/mol) 249.27
MDL Number MFCD00015457
InChI Key RBUSDPZPBBBAKR-UHFFFAOYSA-N
Synonym ethyl 5,6-dimethoxyindole-2-carboxylate, 5,6-dimethoxyindole-2-carboxylic acid ethyl ester, 5,6-dimethoxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5,6-dimethoxy-, ethyl ester, acmc-20aky9, ethyl5,6-dimethoxyindole-2-carboxylate, 1h-indole-2-carboxylicacid, 5,6-dimethoxy-, ethyl ester
PubChem CID 85385
IUPAC Name ethyl 5,6-dimethoxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(OC)=C(OC)C=C2N1

Specifications

Melting Point 175°C to 179°C
Quantity 1 g
Beilstein 211948
Formula Weight 249.27
Percent Purity ≥98%
Chemical Name or Material Ethyl 5,6-dimethoxyindole-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.