accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOIOOH3C

CAS RN 164178-33-0

IUPAC Name: 6-iodo-3-(4-methoxybenzoyl)-2-methyl-1-[2-(morpholin-4-yl)ethyl]-1H-indole
Synonyms: 6-iodo-3-(4-methoxybenzoyl)-2-methyl-1-[2-(morpholin-4-yl)ethyl]indole
Molecular Weight (g/mol): 504.37
Molecular Formula: C23H25IN2O3
InChi Key: JHOTYHDSLIUKCJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=C(C)N(CCN2CCOCC2)C2=CC(I)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results