CAS RN 1642554-49-1
CAS RN 1642554-49-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ ING-2 AM
Fluorescent sodium ion (Na+) indicator, membrane permeable