accessibility menu, dialog, popup

UserName

Viewing profile as user:
FMgCl

CAS RN 1643-73-8

IUPAC Name: chloro[(4-fluorophenyl)methyl]magnesium
Synonyms: chloro[(4-fluorophenyl)methyl]magnesium
Molecular Weight (g/mol): 168.88
Molecular Formula: C7H6ClFMg
InChi Key: PQDGQUPDDGUKLP-UHFFFAOYSA-M
SMILES: FC1=CC=C(C[Mg]Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results