accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NFFFSOOO

CAS RN 164411-06-7

IUPAC Name: N-phenylanilinium trifluoromethanesulfonate
Synonyms: N-phenylanilinium triflate
Molecular Weight (g/mol): 319.3
Molecular Formula: C13H12F3NO3S
InChi Key: MGEGQAUINMTPGX-UHFFFAOYSA-N
SMILES: [O-]S(=O)(=O)C(F)(F)F.[NH2+](C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (3)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results