Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylammonium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. D36835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36835G 5 g
D36831G 1 g
D368325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D36835G Supplier TCI America Supplier No. D36835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 164411-06-7
Molecular Formula C13H12F3NO3S
Molecular Weight (g/mol) 319.298
InChI Key MGEGQAUINMTPGX-UHFFFAOYSA-N
Synonym DPAT
PubChem CID 11209371
IUPAC Name diphenylazanium;trifluoromethanesulfonate
SMILES C1=CC=C(C=C1)[NH2+]C2=CC=CC=C2.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Color White
Quantity 5 g
Formula Weight 319.30
Percent Purity ≥98.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material Diphenylammonium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.