accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFClCl

CAS RN 1649-08-7

IUPAC Name: 1,2-dichloro-1,1-difluoroethane
Synonyms: 1,2-dichloro-1,1-difluoroethane
Molecular Weight (g/mol): 134.93
Molecular Formula: C2H2Cl2F2
InChi Key: SKDFWEPBABSFMG-UHFFFAOYSA-N
SMILES: FC(F)(Cl)CCl
Element 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)