accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16497-41-9

IUPAC Name: N1,N2-diphenylbenzene-1,2-dicarboxamide
Synonyms: N1,N2-diphenylbenzene-1,2-dicarboxamide
Molecular Weight (g/mol): 316.36
Molecular Formula: C20H16N2O2
InChi Key: ZYACJWRVLBPZNG-UHFFFAOYSA-N
SMILES: O=C(NC1=CC=CC=C1)C1=CC=CC=C1C(=O)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results