accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 16533-71-4

IUPAC Name: 4-methyl-3,5-dinitrobenzoic acid
Synonyms: 4-methyl-3,5-dinitrobenzoic acid
Molecular Weight (g/mol): 226.14
Molecular Formula: C8H6N2O6
InChi Key: LZWWZQXBKVZKIP-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1[N+]([O-])=O)C(O)=O)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results