accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 1655-07-8

IUPAC Name: ethyl 2-oxocyclohexane-1-carboxylate
Synonyms: ethyl 2-oxocyclohexane-1-carboxylate
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: FGSGHBPKHFDJOP-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCCC1=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)