Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Carbethoxycyclohexanone, MP Biomedicals

Catalog No. ICN204353 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204353 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204353 Supplier MP Biomedicals, Inc Supplier No. 0520435310

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYNA-N
Synonym ethyl 2-oxocyclohexanecarboxylate, ethyl 2-cyclohexanonecarboxylate, 2-carbethoxycyclohexanone, ethyl cyclohexanone-2-carboxylate, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-oxocyclohexanecarboxylic acid ethyl ester, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O

Specifications

Quantity 10 g
Formula Weight 170.21
Chemical Name or Material 2-Carbethoxycyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.