Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-oxocyclohexanecarboxylate, 95%

Catalog No. AC118331000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC118331000 100 g Glass bottle
AC118330050 5 g Glass bottle
AC118330250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118331000 Supplier Thermo Scientific Chemicals Supplier No. 118331000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1655-07-8 | C9H14O3 | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYNA-N
Synonym ethyl 2-oxocyclohexanecarboxylate, ethyl 2-cyclohexanonecarboxylate, 2-carbethoxycyclohexanone, ethyl cyclohexanone-2-carboxylate, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-oxocyclohexanecarboxylic acid ethyl ester, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O

Specifications

Density 1.0600g/mL
Boiling Point 106.0°C (11.0 mmHg)
Flash Point 85°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (O=)C6H9CO2C2H5
Refractive Index 1.4775 to 1.4795
Quantity 100 g
Beilstein 10, IV, 2606
Specific Gravity 1.06
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material Ethyl 2-oxocyclohexanecarboxylate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-731-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.