accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNBrNOO

CAS RN 165669-16-9

IUPAC Name: 5-bromo-7-nitro-1H-indole
Synonyms: 5-bromo-7-nitro-1H-indole
Molecular Weight (g/mol): 241.04
Molecular Formula: C8H5BrN2O2
InChi Key: WERRDKWLHFDNTO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2NC=CC2=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results