accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOOHOHOBrOSOOBr

CAS RN 16574-43-9

IUPAC Name: 2',7'-dibromo-3',4',5',6'-tetrahydroxyspiro[2,1λ⁶-benzoxathiole-3,9'-xanthene]-1,1-dione
Synonyms: bromopyrogallol red
Molecular Weight (g/mol): 558.15
Molecular Formula: C19H10Br2O8S
InChi Key: QFXYYBXMARTXHI-UHFFFAOYSA-N
SMILES: OC1=C(O)C2=C(C=C1Br)C1(OS(=O)(=O)C3=CC=CC=C13)C1=CC(Br)=C(O)C(O)=C1O2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results