accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFFFNOO

CAS RN 16583-08-7

IUPAC Name: 2,3,4,5-tetrafluoro-6-nitrobenzoate
Synonyms: 2,3,4,5-tetrafluoro-6-nitrobenzoate
Molecular Weight (g/mol): 238.07
Molecular Formula: C7F4NO4
InChi Key: JQGBMYKEWLSLCI-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=C(F)C(F)=C(F)C(F)=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results