accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1660-94-2

IUPAC Name: diethyl [(diethoxyphosphoryl)methyl]phosphonate
Synonyms: diethyl (diethoxyphosphoryl)methylphosphonate
Molecular Weight (g/mol): 288.22
Molecular Formula: C9H22O6P2
InChi Key: STJWVOQLJPNAQL-UHFFFAOYSA-N
SMILES: CCOP(=O)(CP(=O)(OCC)OCC)OCC
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)