CAS RN 166023-29-6
IUPAC Name:
(3R)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Synonyms:
(3R)-1-(tert-butoxy)-5-methyl-1-oxohexan-3-aminium
Molecular Weight (g/mol):
202.32
Molecular Formula:
C11H24NO2
InChi Key:
WBOPVTYXLFEVGP-SECBINFHSA-O
SMILES:
CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C
1
–
1
of
1
results
1
Promotions Available
tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, Thermo Scientific™
tert-Butyl (3R)-3-amino-5-methylhexanoate, 97%, Thermo Scientific™
CAS: 166023-29-6 Molecular Formula: C11H24NO2 Molecular Weight (g/mol): 202.32 MDL Number: MFCD01076257 InChI Key: WBOPVTYXLFEVGP-SECBINFHSA-O PubChem CID: 2733825 IUPAC Name: tert-butyl (3R)-3-amino-5-methylhexanoate SMILES: CC(C)C[C@@H]([NH3+])CC(=O)OC(C)(C)C