accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHOO

CAS RN 16611-84-0

IUPAC Name: 2-hydroxy-6-pentadecylbenzoic acid
Synonyms: anacardic acid
Molecular Weight (g/mol): 348.53
Molecular Formula: C22H36O3
InChi Key: ADFWQBGTDJIESE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC1=CC=CC(O)=C1C(O)=O
Form 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results