accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOH2COCH2OCH2

CAS RN 16646-44-9

IUPAC Name: 3-[1,2,2-tris(prop-2-en-1-yloxy)ethoxy]prop-1-ene
Synonyms: 3-[1,2,2-tris(prop-2-en-1-yloxy)ethoxy]prop-1-ene
Molecular Weight (g/mol): 254.33
Molecular Formula: C14H22O4
InChi Key: BXAAQNFGSQKPDZ-UHFFFAOYSA-N
SMILES: C=CCOC(OCC=C)C(OCC=C)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results