Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraallyloxyethane, TCI America™
SDP

Catalog No. T0030500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0030500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0030500ML Supplier TCI America Supplier No. T0030500ML

Specifications

CAS 16646-44-9
Color Yellow
Boiling Point 160°C
Molecular Formula C14H22O4
MDL Number MFCD00043927
Quantity 500 mL
Synonym 1,1,2,2-Tetrakis(allyloxy)ethane
InChI Key BXAAQNFGSQKPDZ-UHFFFAOYSA-N
SMILES C=CCOC(OCC=C)C(OCC=C)OCC=C
IUPAC Name 3-[1,2,2-tris(prop-2-en-1-yloxy)ethoxy]prop-1-ene
Molecular Weight (g/mol) 254.33
PubChem CID 85528
Formula Weight 254.33
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraallyloxyethane
Show More Show Less
EINECSNumber (2)-2460
RTECSNumber KI8925000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.