accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1667-99-8

IUPAC Name: trisodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichloro-3-sulfonatophenyl)methyl}-2-hydroxy-3-methylbenzoate
Synonyms: trisodium 5-{[(1E)-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene](2,6-dichloro-3-sulfonatophenyl)methyl}-2-hydroxy-3-methylbenzoate
Molecular Weight (g/mol): 605.28
Molecular Formula: C23H13Cl2Na3O9S
InChi Key: FUIZKNBTOOKONL-DPSBJRLESA-K
SMILES: [Na+].[Na+].[Na+].CC1=CC(=CC(C([O-])=O)=C1O)C(=C1\C=C(C)C(=O)C(=C1)C([O-])=O)\C1=C(Cl)C=CC(=C1Cl)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (8)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (5)
  • (2)
Color 
  • (2)
  • (4)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)