Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chrome Azurol S

Catalog No. AC190050250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190050250 25 g Glass bottle
AC190051000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190050250 Supplier Thermo Scientific Chemicals Supplier No. 190050250
  • $136.00
    $98.65 / Each of 1

    Save $37.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 1667-99-8 | C23H13Cl2Na3O9S | 605.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym C.I. 43825, Mordant Blue 29
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

Infrared Spectrum Authentic
Lambda Max 424 to 436nm (c=0.03g/L in H2O)
Packaging Glass bottle
UV Spectrometry Complies
Color Black to Brown
Quantity 25 g
Physical Form Crystalline Powder
Formula Weight 605.29
Chemical Name or Material Chrome Azurol S
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 216-787-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.