Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mordant Blue 29, TCI America™
SDP

Catalog No. C09795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C09795G Supplier TCI America Supplier No. C09795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym mordant blue 29, chromeazurol s, chrome azurol s, trisodium 5-3-carboxylato-5-methyl-4-oxo-cyclohexa-2,5-dien-1-ylidene-2,6-dichloro-3-sulfonato-phenyl methyl-2-hydroxy-3-methyl-benzoate, trisodium 5-3-carboxylato-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene-2,6-dichloro-3-sulfonatophenyl methyl-2-hydroxy-3-methylbenzoate
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

Quantity 5 g
Formula Weight 605.28
Physical Form Crystalline Powder
Chemical Name or Material Mordant Blue 29
EINECSNumber (5)-2088
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.