CAS RN 16674-78-5
Magnesium acetate tetrahydrate, ACS, 98.0-102.0%
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate Solution, 354g/L (40.1 g/L as Magnesium, for AOAC Method 961.08), Fisher Chemical
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium acetate tetrahydrate, 98% (metals basis)
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium;diacetate;tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate, Tetrahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Thermo Scientific Chemicals Magnesium acetate tetrahydrate, for biochemistry
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium;diacetate;tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate Tetrahydrate, ≥99%, Reagent Grade, Solstice
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium acetate tetrahydrate, 97.5%, extra pure, crystalline
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium acetate tetrahydrate, Puratronic™, 99.997% (metals basis)
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium acetate tetrahydrate, 99.5%, for analysis
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium acetate tetrahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate Tetrahydrate, Solstice
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium;diacetate;tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate, Promega
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
Magnesium Acetate Tetrahydrate, Fisher BioReagents
CAS: 16674-78-5 | C4H14MgO8 | 214.45 g/mol
Magnesium Acetate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical
CAS: 16674-78-5 Molecular Formula: C4H14MgO8 Molecular Weight (g/mol): 214.45 MDL Number: MFCD00149214 InChI Key: XKPKPGCRSHFTKM-UHFFFAOYSA-L Synonym: magnesium acetate tetrahydrate,cromosa,acetic acid, magnesium salt, tetrahydrate,unii-i01g0ejc3b,i01g0ejc3b,magnesium diacetate tetrahydrate,magnesium diethanoate tetrahydrate,acmc-20aklu,magnesiumacetatetetrahydrate,ksc497c8p PubChem CID: 134717 IUPAC Name: magnesium(2+) diacetate tetrahydrate SMILES: O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O
- 1
- 2