accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSSOOCH3O

CAS RN 1668554-22-0

IUPAC Name: 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene-5,11-dicarbaldehyde
Synonyms: 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene-5,11-dicarbaldehyde
Molecular Weight (g/mol): 502.73
Molecular Formula: C28H38O4S2
InChi Key: JDPODAXHWNAIOO-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C2C=C(SC2=C(OCCCCCCCC)C2=C1SC(C=O)=C2)C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results