accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOH3C

CAS RN 16691-79-5

IUPAC Name: 1,3-dimethyl 2-oxo-4,5-diphenylcyclopenta-1(5),3-diene-1,3-dicarboxylate
Synonyms: 1,3-dimethyl 2-oxo-4,5-diphenylcyclopenta-1(5),3-diene-1,3-dicarboxylate
Molecular Weight (g/mol): 348.35
Molecular Formula: C21H16O5
InChi Key: NTXFYWYVVGMAGT-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C(=C(C(=O)OC)C1=O)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results