CAS RN 167144-80-1
IUPAC Name:
5-(2-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}ethyl)-1-methylpyrrolidin-3-ol hydrochloride
Synonyms:
5-(2-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}ethyl)-1-methylpyrrolidin-3-ol hydrochloride
Molecular Weight (g/mol):
391.94
Molecular Formula:
C22H30ClNO3
InChi Key:
OUKRSTJUAQEATQ-UHFFFAOYNA-N
SMILES:
Cl.COC1=CC=CC(CCC2=CC=CC=C2OCCC2CC(O)CN2C)=C1
1
–
1
of
1
results
1
R&D Systems™ R-96544 hydrochloride
CAS: 167144-80-1 Molecular Formula: C22H30ClNO3