accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2ONHN

CAS RN 1672-50-0

IUPAC Name: 5,6-diamino-1,4-dihydropyrimidin-4-one
Synonyms: 5,6-diamino-1H-pyrimidin-4-one
Molecular Weight (g/mol): 126.12
Molecular Formula: C4H6N4O
InChi Key: PWRHKLKFADDKHS-UHFFFAOYSA-N
SMILES: NC1=C(N)C(=O)N=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results