Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Diamino-6-hydroxypyrimidine 96.0+%, TCI America™
SDP

Catalog No. D4488200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4488200MG 200 mg
D44881G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4488200MG Supplier TCI America Supplier No. D4488200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1672-50-0
Molecular Formula C4H6N4O
Molecular Weight (g/mol) 126.12
MDL Number MFCD00006114
InChI Key PWRHKLKFADDKHS-UHFFFAOYSA-N
Synonym 5,6-Diamino-4(1H)-pyrimidinone, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4-hydroxypyrimidine
PubChem CID 74287
IUPAC Name 5,6-diamino-1,4-dihydropyrimidin-4-one
SMILES NC1=C(N)C(=O)N=CN1

Specifications

Melting Point 244°C
Quantity 200 mg
Formula Weight 126.12
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Diamino-6-hydroxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.