CAS RN 167416-28-6
CAS RN 167416-28-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
606.64
Molecular Formula:
C36H56FeP2
InChi Key:
LVJMXQBMWBNENE-UHFFFAOYNA-N
SMILES:
[Fe].[CH]1[CH][CH][CH][CH]1.CC([C]1[CH][CH][CH][C]1P(C1CCCCC1)C1CCCCC1)P(C1CCCCC1)C1CCCCC1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich ™-1-[(Sp)-2-(Dicyclohexylphosphino)Ferrocenylethyl]Diphenylphosphine
Sigma Aldrich ™-1-[(Sp)-2-(Dicyclohexylphosphino)Ferrocenylethyl]Diphenylphosphine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.